Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LDL R, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 11 publications

Manufacturer:  Novus Biologicals NBP106709

 View more versions of this product

Catalog No. NBP106709


Add to cart

Description

Description

LDLR Polyclonal antibody specifically detects LDLR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

LDLR
1.5 mg/ml
Western Blot 0.5 ug/ml, Simple Western 1:100, Immunohistochemistry 1:1000 - 1:3000, Immunocytochemistry/Immunofluorescence 1:1000-1:10000, Immunohistochemistry-Paraffin 1:1000 - 1:3000
P01130
FH, FHC, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3
Rabbit
IgG
0.1 ml
Cancer, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction
Primary
3949
Human, Mouse
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS with 0.1% Sodium Azide
Affinity Purified
LDLR
Synthetic peptide made to an internal portion of the human LDL Receptor protein (within residues 500-550). [Swiss-Prot# P01130]
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This is specific for both the unglycosylated and glycosylated forms of the LDL Receptor.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only