Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEDGF (Lens Epithelium-Derived Growth Factor) Mouse anti-Human, Unlabeled, Clone: 26, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611715

 View more versions of this product

Catalog No. BDB611715


Add to cart

Description

Description

p52/p75 proteins were first identified through their interaction with transcriptional factors and splicing factors. p75 is encoded by exons 1-15 and contains 530 amino acids, while the alternatively spliced p52 is encoded by exons 1-9 and contains 333 amino acids. Both p52 and p75 have strong interactions with the VP16 activation domain and with various components of the general transcriptional machinery. p52 is the more active transcription activator and has also been shown to recruit the pre-mRNA splicing factor ASF/SF to sites of active transcription. Interestingly, p52/p75 has also been identified as Lens Epithelium-Derived Growth Factor (LEDGF). In serum free media, LEDGF stimulates the growth and survival of a variety of cells, such as lens epithelial cells, retinal photoreceptor cells, COS7 cells, skin fibroblasts, and keratinocytes. In retinal photoreceptor cells, LEDGF is found in the nucleus where it promotes resistance to serum starvation and thermal stress. Thus, the LEDGF gene encodes splice variants that may regulate gene transcription and mRNA splicing of a variety of proteins that are essential for cell growth and survival.

Host Species: Mouse
Clone: 26
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Human LEDGF aa. 85-188

Immunofluorescence, Western Blotting

Specifications

Specifications

LEDGF (Lens Epithelium-Derived Growth Factor)
26
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
In vitro Assay, Western Blot
250μg/mL
Lens Epithelium-Derived Growth Factor
Affinity Purified
Human LEDGF aa. 85-188
150μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.