Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEF1 Mouse anti-Human, Mouse, Clone: REMB6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112421

Catalog No. 01-674-298


Add to cart

Description

Description

A suggested positive control for this product is Jurkat cell lysate.

Lymphoid Enhancer Factor -1 is a transcription factor of the High Mobility Group of DNA binding proteins. It is one member of a family of four proteins referred to as LEF/TCF transcription factors (LEF-1, TCF-1, TCF-3 and TCF-4). These factors play a crucial role in WNT/WIngless signaling, a signal transduction cascade that directs cell differentiation. Aberrant activation of the WNT/Wingless pathway is also a root cause in the genesis of certain cancers such as colon cancer, melanoma and breast cancer.
Specifications

Specifications

LEF1
Monoclonal
1 mg/mL
PBS with 0.08% sodium azide
Q9UJU2, P27782
HMG Binding Domain
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
16842, 51176
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
REMB6
Unconjugated
LEF1
Liquid
LEF1
Full-length recombit LEF-1 protein.
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit