Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEO1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511820

Catalog No. PIPA511820

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The PAF1 complex is a multifunctional complex. The PAF1 complex interacts with POLR2A. It may be involved in both initiation and elongation, histone methylation and RNA processing. Overexpression of LEO1 induces cell growth arrest and premature senescence of fibroblasts.
Specifications

Specifications

LEO1
Polyclonal
PBS with 0.09% sodium azide
Q8WVC0
RDL; RNA polymerase-associated protein LEO1; Replicative senescence down-regulated leo1-like protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123169
Western Blot
Unconjugated
LEO1
Liquid
LEO1
KLH conjugated synthetic peptide between 130-159 amino acids from the N-terminal region of human LEO1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit