Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Leptin Oligoclonal, Invitrogen™

LeptinOligoclonal antibody 710145 from Thermo Scientific™ Pierce™ specifically detects Leptin in Rat samples, and it is validated for WB

Manufacturer:  Invitrogen 710145

Encompass

Catalog No. 71-014-5


Add to cart

Description

Description

Leptin is a 167-amino acid peptide hormone encoded by the obesity gene (OB) and secreted by adipocytes. It is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, and influences pubertal development and fetal growth; it is involved in obesity, anorexia nervosa, insulin resistance, and hypertension. Serum leptin levels correlate with body fat amount.

710145 detects Leptin in Rat samples. 710145 has been successfully used in WB procedures. The immunogen for 710145 is recombinant protein corresponding to amino acids 26-59 of rat leptin.

Applications:

Western Blotting

Specifications

Specifications

Leptin
Unconjugated
P50596
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Polyclonal
Rat
1HCLC
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
Recombinant protein corresponding to amino acids 26-59 of rat leptin
Protein A
Primary
25608
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit