Learn More
Leptospira biflexa Rabbit anti-Bacteria, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific PA173008
Catalog No. PIPA173008
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
The PA1-73008 antibody reacts with Leptospira biflexa. PA1-73008 has been successfully used in immunofluorescence applications. The PA1-73008 antibody was raised against whole L. biflexa organism. Recommended short term storage, less than 6 months, is liquid at 4°C under subdued light. For longer term storage, aliquot and freeze.
Leptospirosis is a contagious disease affecting both animals and humans, spread by infection with a bacterial pathogen called Leptospira. Infection may result in chronic liver and kidney disease. Leptospires are known as "aquatic spirochetes": they thrive in water and appear long and helical with a characteristic hook on one or both ends. These organisms are divided into two species, Leptospira biflexa and Leptospira interogans.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Leptospira biflexa
Polyclonal
FITC
Rabbit
Purified
RUO
Bacteria
Antibody
IgG
Immunofluorescence
4-5 mg/mL
L. biflexa; Leptospirosis; Leptospira
Whole L. biflexa organism.
1 mL
Primary
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Liquid
Recommendations
Recommendations