Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leptospira biflexa, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128914

Catalog No. PIPA128914

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Leptospirosis is a contagious disease affecting both animals and humans, spread by infection with a bacterial pathogen called Leptospira. Infection may result in chronic liver and kidney disease. Leptospires are known as aquatic spirochetes: they thrive in water and appear long and helical with a characteristic hook on one or both ends. These organisms are divided into two species, Leptospira biflexa and Leptospira interogans.

PA1-28914 detects Leptospira biflexa from bacteria samples.PA1-28914 has been successfully used in immunofluorescence applications.The PA1-28914 immunogen is: Tissue / cell preparation (Whole organism).
Specifications

Specifications

Leptospira biflexa
Polyclonal
Unconjugated
Purified
Rabbit
IgG
100μL
Store at -20°C
Primary
Bacteria
Immunocytochemistry, Immunofluorescence
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
L. biflexa, Leptospirosis, Leptospira
Tissue/cell preparation (Whole organism).
Protein A
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit