Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRG1 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523266

Catalog No. PIPA523266


Add to cart

Description

Description

LRG1 is a 347 amino acid containing leucine-rich secreted protein, belonging to leucine-rich repeat (LRR) family of proteins and contains eight LRR (leucine rich) repeats, a galactosamine and four glucosamine oligosaccharides attached and has two intrachain disulfide bonds. LRG1, a regulatory protein, is usually found in plasma and is expressed during granulocyte differentiation. Different functions of LRG1 include protein-protein interaction, signal transduction, cell adhesion and development. LRG1 expression is detected in neutrophil fraction of bone marrow and peripheral blood and in the bone marrow mononuclear cell fraction. LRG1 expression is usually upregulated in early DMSO-induced granulocyte differentiation of human promyelocytic leukemia HL-60 cells and also in GCSF-treated human cells derived from a patient with myeloproliferative disorder. Thus LRG1 acts as a novel marker for early neutrophilic granulocyte differentiation.
Specifications

Specifications

LRG1
Polyclonal
Unconjugated
LRG1
Liquid
LRG1
Synthetic peptide (311 YRWLQAQKDKMFSQNDTR 328) of Leucine rich alpha 2 glycoprotein 1 (LRG1) protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Non-human primate
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P02750
HMFT1766, LRG, 1300008B03Rik, 2310031E04Rik, leucine rich alpha 2 glycoprotein, leucine-rich alpha-2-glycoprotein
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
116844, 76905
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit