Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Leucyl-cystinyl Aminopeptidase/LNPEP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232111

Catalog No. NBP232111


Add to cart

Description

Description

Leucyl-cystinyl Aminopeptidase/LNPEP Polyclonal antibody specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Leucyl-cystinyl Aminopeptidase/LNPEP
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
NP_005566.2
angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
LNPEP
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Oxytocinase using the numbering given in entry NP_005566.2 (GeneID 4012).
Immunogen affinity purified
RUO
Primary
4012
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only