Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LTA4H Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528216

Catalog No. PIPA528216


Add to cart

Description

Description

PA5-28216 targets Leukotriene A4 hydrolase in WB applications and shows reactivity with Human samples. The PA5-28216 immunogen is recombit fragment corresponding to a region within amino acids 1 and 315 of Human Leukotriene A4 hydrolase. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Hydrolyzes an epoxide moiety of leukotriene A4 (LTA-4) to form leukotriene B4 (LTB-4). The enzyme also has some peptidase activity.
Specifications

Specifications

LTA4H
Polyclonal
Unconjugated
LTA4H
Liquid
LTA4H
Recombit fragment corresponding to a region within amino acids 1 and 315 of Human Leukotriene A4 hydrolase
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.86 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P09960
LTA-4 hydrolase, leukotriene A-4 hydrolase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
4048
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit