Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human LFNG protein.

Manufacturer:  Abnova Corporation H00003955B01

Catalog No. 89-005-697


Add to cart

Description

Description

This gene encodes a member of the glycosyltransferase superfamily. The encoded protein is a single-pass type II Golgi membrane protein that functions as a fucose-specific glycosyltransferase, adding an N-acetylglucosamine to the fucose residue of a group of signaling receptors involved in regulating cell fate decisions during development. Mutations in this gene have been associated with autosomal recessive spondylocostal dysostosis 3. Alternatively spliced transcript variants that encode different isoforms have been described, however, not all variants have been fully characterized. [provided by RefSeq

Sequence: MTPGRCCLAADIQVETFIFTDGEDEALARHTGNVVITNCSAAHSRQALSCKMAVEYDRFIESGRKWFCHVDDDNYVNLRALLRLLASYPHTRDVYVGKPSLDRPIQAMERVSENKVRPVHFWFATGGAGFCISRGLALKMSPWASGGHFMNTAERIRLPDDCTIGYIVEALLGVPLIRSGLFHSHLENLQQVPTSELHEQVTLSYGMFENKRNAVHVKGPFSVEADPSRFRSIHCHLYPDTPWCPRTAIF
Specifications

Specifications

LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Unconjugated
No additive
BC014851
SCDO3
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3955
Western Blot
Mouse polyclonal antibody raised against a full-length human LFNG protein.
LFNG
Serum
LFNG
LFNG (AAH14851, 1 a.a. ~ 250 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit