Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALS9B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523573

Catalog No. PIPA523573

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

LGALS9B
Polyclonal
PBS with 0.09% sodium azide
Q3B8N2
gal-9B, galectin-9 like, galectin-9 pseudogene, galectin-9-like protein A, galectin-9B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284194
Flow Cytometry, Western Blot
Unconjugated
LGALS9B
Liquid
LGALS9B
KLH conjugated synthetic peptide between 160-188 amino acids from the central region of human LGALS9B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit