Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520492

Catalog No. PIPA520492


Add to cart

Description

Description

A suggested positive control is 293 cell lysate. PA5-20492 can be used with blocking peptide PEP-0612.

The leucine-rich, glioma inactivated gene 2 (LGI2) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodomit lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predomit auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI2 have not been found to be associated with ADTLE.
Specifications

Specifications

LGI2
Polyclonal
Unconjugated
LGI2
Liquid
LGI2
a 11 amino acid peptide from near the amino terminus of human LGI2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8N0V4, Q8K4Z0
mKIAA1916; leucine-rich glioma-inactivated protein 2; leucine-rich repeat LGI family member 2
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
246316, 305417, 55203
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit