Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520434

Catalog No. PIPA520434


Add to cart

Description

Description

A suggested positive control is MDA-MB-361 cell lysate. PA5-20434 can be used with blocking peptide PEP-0551.

Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
Specifications

Specifications

LGP2
Polyclonal
Unconjugated
DHX58
Liquid
DHX58
A 14 amino acid peptide from near the amino terminus of human LGP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q96C10, Q99J87
D11LGP2, D11lgp2e, LGP2; RIG-I-like receptor LGP2; RLR; RNA helicase LGP2; ortholog of mouse D11lgp2; probable ATP-dependent RNA helicase DHX58; probable ATP-dependent helicase LGP2; protein D11Lgp2 homolog
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
303538, 79132, 80861
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit