Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lgr5/GPR49, Alexa Fluor 405, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128904AF405

Catalog No. NBP128904W


Add to cart

Description

Description

Lgr5/GPR49 Polyclonal antibody specifically detects Lgr5/GPR49 in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, ICC/IF (Negative).
Specifications

Specifications

Lgr5/GPR49
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
LGR5
Synthetic peptide made to an internal portion of the human GPR49/LGR5 protein (within residues 650-700). [Swiss-Prot: O75473]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, Porcine
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, ICC/IF (Negative)
O75473
FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Rabbit
IgG
0.1 ml
Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary
8549
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only