Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgr5/GPR49 Rabbit anti-Human, Mouse, Porcine, DyLight 650, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP128904C
Description
Lgr5/GPR49 Polyclonal antibody specifically detects Lgr5/GPR49 in Human, Mouse, Porcine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Lgr5/GPR49 | |
DyLight 650 | |
Affinity Purified | |
O75473 | |
8549 | |
Rabbit | |
IgG | |
Primary | |
0.1 ml | |
Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway | |
Human,Mouse,Porcine |
Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen) | |
Western Blot, Flow Cytometry, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
LGR5 | |
Synthetic peptide made to an internal portion of the human GPR49/LGR5 protein (within residues 650-700). [Swiss-Prot: O75473] | |
Polyclonal | |
Immunogen affinity purified | |
RUO | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only