Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Lgr6, FITC, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP149696

Catalog No. NBP149696


Add to cart

Description

Description

Lgr6 Polyclonal antibody specifically detects Lgr6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Lgr6
FITC
Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide
Affinity Purified
LGR6
Synthetic peptides corresponding to unique epitope on Lgr6. The peptide sequence corresponding to amino acids 860-915 of the human Lgr6 protein, the peptide was post-synthetically modified to achieve desired antigenicity before coupling to a carrier protein (CAI15929).
115 kDa
0.05 mg
GPCR
Primary
59352
Human, Mouse, Rat
ELISA, Western Blot
Western Blot 1:100-1:2000, ELISA < 1:10000
Q9HBX8
FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody detects a fuzzy band of approximately 115 kDa in PC-Lgr6 samples and few additional bands between MW45-60kDa were also observed. The identity of these bands are not known but could be proteolytic fragment of the full-length Lgr6 molecule.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only