Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHFPL5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523919

Catalog No. PIPA523919


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

This gene is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes. Mutations in this gene result in deafness in humans, and a mutation in a similar gene in mice results in deafness and vestibular dysfunction with severe degeneration of the organ of Corti. It is proposed to function in hair bundle morphogenesis.
Specifications

Specifications

LHFPL5
Polyclonal
PBS with 0.09% sodium azide
Q8TAF8, Q4KL25
DFNB67, TMHS, dJ510O8.8, LHFP-like protein 5, lipoma HMGIC fusion partner-like 5 protein, tetraspan membrane protein of hair cell stereocilia
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
222662, 328789
Western Blot
Unconjugated
LHFPL5
Liquid
LHFPL5
KLH conjugated synthetic peptide between 186-214 amino acids from the C-terminal region of human LHFPL5
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit