Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHFPL2 Rabbit anti-Hamster, Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525166

Catalog No. PIPA525166

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

LHPL2 is a member of the lipoma HMGIC fusion partner (LHFP) gene family, which is a subset of the superfamily of tetraspan transmembrane protein encoding genes.
Specifications

Specifications

LHFPL2
Polyclonal
PBS with 0.09% sodium azide
Q6ZUX7
LHFP-like protein 2, lipoma HMGIC fusion partner-like 2 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100755063, 10184
Western Blot
Unconjugated
LHFPL2
Liquid
LHFPL2
KLH conjugated synthetic peptide between 196-223 amino acids from the C-terminal region of human LHPL2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit