Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LHX3 Rabbit anti-Amphibian, Fish, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA129491

Catalog No. PA129491


Add to cart

Description

Description

PA1-29491 detects LHX3 from Amphibian, mouse, human, rat, fish samples. PA1-29491 has been successfully used in immunocytochemistry, immunofluorescence, immunohistochemistry (paraffin & frozen tissue), Western blot, immunoprecipitation applications. Predicted molecular weight of 44 kDa. The PA1-29491 immunogen is: C-terminal portion of mouse LHX3 protein. Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20° C or below, avoiding multiple freeze-thaw cycles.

This gene encodes a member a large protein family which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein is a transcription factor that is required for pituitary development and motor neuron specification. Mutations in this gene have been associated with a syndrome of combined pituitary hormone deficiency and rigid cervical spine. Two transcripts variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

LHX3
Polyclonal
Unconjugated
LHX3
Liquid
LHX3
C-terminal portion of mouse LHX3 protein.
Protein A
RUO
Antibody
Polyclonal
Amphibian, Fish, Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Q9UBR4, P50481
CPHD3, DKFZp762A2013, LIM3, M2-LHX3
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
16871, 170671, 8022
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit