Learn More
anti-Lhx8, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP132840
Catalog No. NBP132840
Add to cart
Description
Description
Lhx8 Polyclonal antibody specifically detects Lhx8 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
Specifications
Lhx8
1.0mg/mL
Western Blot 1:1000-1:10000, Immunoprecipitation 1:100-1:500
Affinity Purified
LHX8
Full length human LHX8 Recombinant protein.
39 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot, Immunoprecipitation
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
LIM homeobox 8, LIM homeobox protein 8
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
431707
Human