Learn More
Lidocaine Mouse anti-Chemical, Clone: LC12-402.3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA110755
Catalog No. 01-671-985
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
MA1-10755 detects Lidocaine. MA1-10755 has been successfully used in ELISA applications. The MA1-10755 immunogen is Lidocaine, conjugated to KLH.
Lidocaine is a synthetic aminoethylamide with local anesthetic and antiarrhythmic properties. Lidocaine stabilizes the neuronal membrane by binding to and inhibiting voltage-gated sodium channels, thereby inhibiting the ionic fluxes required for the initiation and conduction of impulses and effecting local anesthesia. [National Center for Biotechnology Information. PubChem Compound Database; CID=3676]
Specifications
Specifications
Lidocaine
Monoclonal
∽1 mg/mL
PBS with no preservative; pH 7.4
Lignocaine; Xylocaine; Lidoderm; Anestacon; C14H22N2O
Lidocaine, conjugated to KLH.
Purified
RUO
Antibody
Monoclonal
ELISA
LC12-402.3
Unconjugated
Liquid
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
Chemical