Learn More
Light Chain, lambda Mouse anti-Human, Brilliant Violet 510, Clone: JDC-12, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 563537
Catalog No. BDB563537
Add to cart
Description
Description
The JDC-12 monoclonal antibody specifically binds to human immunoglobulin light chain, lambda ( λ ). It does not not bind to immunoglobulin κ light chains or heavy chains.
Specifications
Specifications
Light Chain, lambda
JDC-12
Ig λ; IGL@; IGL; Immunoglobulin lambda locus
Affinity Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
Brilliant Violet 510
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal