Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIMD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524973

Catalog No. PIPA524973


Add to cart

Description

Description

This antibody is predicted to react with chicken based on sequence homology.

Specifications

Specifications

LIMD2
Polyclonal
PBS with 0.09% sodium azide
Q9BT23
LIM domain-containing protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
80774
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
LIMD2
Liquid
LIMD2
KLH conjugated synthetic peptide between 84-113 amino acids from the N-terminal region of human LIMD2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit