Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lin-28 Mouse anti-Human, Alexa Fluor 647, Clone: 6D1F9, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564280

Catalog No. BDB564280

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Host Species: Mouse
Clone: 6D1F9
Isotype: IgG1, κ
Species Reactivity: Human
Immunogen: Internal region human Lin-28

Intracellular staining, Flow Cytometry

Specifications

Specifications

Lin-28
6D1F9
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Murine
50 Tests
Primary
Human
Flow Cytometry, Immunofluorescence, In vitro Assay
Alexa Fluor 647
Affinity Purified
IgG1, κ
RUO
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit