Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN28A Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523408

Catalog No. PIPA523408


Add to cart

Description

Description

Lin-28 homolog A is a protein that in humans is encoded by the LIN28 gene. It is a marker of undifferentiated human embryonic stem cells and has been used to enhance the efficiency of the formation of induced pluripotent stem (iPS) cells from human fibroblasts. It encodes a cytoplasmic mRNA-binding protein that binds to and enhances the translation of the Igf2 mRNA. LIN28 has also been shown to bind to the let-7 pre-miRNA and block production of the mature let-7 microRNA in mouse embryonic stem cells.
Specifications

Specifications

LIN28A
Polyclonal
Unconjugated
LIN28A
Liquid
LIN28A
Synthetic peptide (117 CIGSERRPKGKSMQKRRSK 135) of LIN28 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q9H9Z2, Q8K3Y3
CSDD1, LIN-28, LIN28, ZCCHC1, lin-28A, RNA-binding protein LIN-28, protein lin-28 homolog A, zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
500562, 79727, 83557
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit