Learn More
Lineage Antibody Cocktail, with Isotype Control Mouse anti-Mouse, PerCP-Cy5.5, Clone: 145-2C11, M1/70, RA3-6B2, TER-119, RB6-8C5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 561317
Description
The PerCP-Cy™5.5 Mouse Lineage Antibody Cocktail has been designed to react with cells from the major hematopoietic lineages, such as T lymphocytes, B lymphocytes, monocytes/macrophages, NK cells, erythrocytes, and granulocytes. This pre-diluted Cocktail of five PerCP-Cy™5.5-conjugated antibodies is designed to label lineage marker-positive cells for exclusion to facilitate the flow cytometric identification of lineage marker-negative hematopoietic progenitors in mouse bone marrow. Components include clone 145-2C11, which recognizes Mouse CD3e; M1/70, whichrecognizes CD11b; RA3-6B2, which recognizes CD45R/B220; TER-119, which recognizes Ly-76, mouse erythroid cells; and RB6-8C5, which recognizes Ly-6G and Ly-6C. PerCP-Cy™5.5 Mouse Lineage Isotype Control Cocktail contains equivalent concentrations of isotype-matched negative-control immunoglobulin. Additional fluorochrome-labeled reagents may be combined with the PerCP-Cy™5.5 Mouse Lineage Antibody Cocktail, and the PerCP-Cy™5.5 Mouse Lineage Isotype Control Cocktail, to further characterize hematopoietic progenitor subpopulations.
Specifications
Lineage Antibody Cocktail, with Isotype Control | |
Monoclonal | |
PerCP-Cyanine5.5 | |
Affinity Purified | |
RUO | |
PerCP-Cy 5.5 Mouse Lineage Antibody Cocktail (100 tests), PerCP-Cy5.5 Mouse Lineage Isotype Control Cocktail (100 tests) | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
145-2C11, M1/70, RA3-6B2, TER-119, RB6-8C5 | |
Mouse | |
100 Tests | |
Primary | |
Mouse |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.