Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LINGO1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525941

Catalog No. PIPA525941

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse and non-human primate based on sequence homology.

LINGO1 is a functional component of the Nogo receptor signaling complex(RTN4R/NGFR) in RhoA activation responsible for some inhibition of axonal regeneration by myelin-associated factors. It is also an important negative regulator of oligodentrocyte differentiation and axonal myelination.
Specifications

Specifications

LINGO1
Polyclonal
PBS with 0.09% sodium azide
Q96FE5, Q9D1T0
LERN1, LRRN6A, UNQ201, leucine rich repeat neuronal 6A, leucine-rich repeat and immunoglobilin domain-containing protein 1, leucine-rich repeat and immunoglobulin-like domain-containing nogo receptor-interacting protein 1, leucine-rich repeat neuronal protein 1, leucine-rich repeat
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
235402, 84894
Western Blot
Unconjugated
LINGO1
Liquid
LINGO1
KLH conjugated synthetic peptide between 57-85 amino acids from the N-terminal region of human LINGO1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit