Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipid A Mouse anti-Bacteria, Clone: 26-5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA135585

Catalog No. 01-671-023

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

LPS is chemically composed of a lipid part, which is responsible for the endotoxic activities of LPS, and a heteropolysaccharide which generally consists of an oligosaccharide, termed the core region, and, in some bacterial families, of the so-called O chain, composed of repeating oligosaccharide units. Lipid A is the glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria.

Specifications

Specifications

Lipid A
Monoclonal
0.1 mg/mL
Assay Dependent
Liquid
Mouse
IgG2b
5 mL
4° C
Primary
Bacteria
ELISA
26-5
Unconjugated
PBS with 0.1% BSA and 0.02% sodium azide
Lipid A free
Synthetic lipid A.
Purified
RUO
Antibody
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit