Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lipid A LPS, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA186403

Catalog No. PA186403

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

LPS is chemically composed of a lipid part, which is responsible for the endotoxic activities of LPS, and a heteropolysaccharide which generally consists of an oligosaccharide, termed the core region, and, in some bacterial families, of the so-called O chain, composed of repeating oligosaccharide units. Lipid A is the glucosamine-based phospholipid which forms part of the bacterial outer membrane and anchors LPS to the surface of the bacteria.

This antibody cross-reacts with P. aeruginosa, K. pneumoniae, S. enteriditis, E. aerogenes, E. hermanii, Y. enterocolitica and S. sonnei.
Specifications

Specifications

Lipid A LPS
Polyclonal
Unconjugated
Purified
Goat
IgG
1mL
Store at 4°C or Store at -20°C if preferred
Primary
Bacteria
Immunofluorescence
4mg/mL
PBS with 0.1% sodium azide
Lipid A Lipopolysaccharide
Whole cell preparation of lipid A from E.coli O157.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit