Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lipin 1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11057150G
Description
Lipin 1 Polyclonal specifically detects Lipin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Lipin 1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q14693 | |
LPIN1 | |
A synthetic peptide to an internal region (within residues 300-400) of the human LPIN1 protein. [Swiss-Prot# Q14693] | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DKFZp781P1796, EC 3.1.3.18, EC 3.1.3.4, EC 5.4.2.7, KIAA0188phosphatidate phosphatase LPIN1, lipin 1, lipin-1, PAP1 | |
Rabbit | |
97 kDa | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction | |
23175 | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction