Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria monocytogenes, Polyclonal, Invitrogen™

Catalog No. PIPA185649
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
PIPA185649 1 mL
1 options

Catalog No. PIPA185649

Supplier: Thermo Scientific PA185649

Rabbit Polyclonal Antibody

The genus Listeria comprises six species: L. monocytogenes, L. innocua, L. welshimeri, L. seeligeri, L. ivanovii and L. grayi. Listeria monocytogenes, the most commonly isolated pathogenic member, is associated with a wide spectrum of human and animal diseases. In the smear from the original tissue, L. monocytogenes may appear as gram-positive coccobacilli that may be confused with Streptococcus agalactiae (group B), enterococci, or Corynebacterium spp. Listeria is differentiated from streptococci by a positive catalase test.

This product requires protein digestion pre-treatment of paraffin sections using trypsin or pronase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Listeria monocytogenes
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 4mg/mL
Conjugate Unconjugated
Gene Alias L. monocytogenes
Host Species Rabbit
Immunogen Listeria monocytogenes; ATCC strain 43251
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C or Store at -20°C if preferred
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.