Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Listeria Rabbit anti-Bacteria, FITC, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA173128

Catalog No. PIPA173128


Add to cart

Description

Description

The PA1-73128 antibody reacts with Listeria species. PA1-73128 has been successfully used in immunofluorescence applications. Acetone fixation is recommended prior to staining. The PA1-73128 antibody was raised against Listeria monocytogenes; ATCC strain 43251. Product can be stored at 4°C for up to 6 months. For long term storage, please aliquot and store at -20°C, avoiding freeze/thaw cycles. Store in the dark.

Specifications

Specifications

Listeria
Polyclonal
FITC
Liquid
Listeria monocytogenes; ATCC strain 43251.
Purified
RUO
Antibody
Polyclonal
Immunofluorescence
4-5 mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit