Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginase 1 Rabbit anti-Bovine, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126408

Catalog No. PIPA126408


Add to cart

Description

Description

PA1-26408 detects liver Arginase from bovine samples. PA1-26408 has been successfully used in ELISA, IM and Western blot procedures. The PA1-26408 immunogen is Arginase from bovine liver.

Arginase catalyzes the hydrolysis of arginine to ornithine and urea. At least two isoforms of mammalian arginase exist (types I and II) which differ in their tissue distribution, subcellular localization, immunologic crossreactivity and physiologic function. The type I isoform encoded by this gene, is a cytosolic enzyme and expressed predomitly in the liver as a component of the urea cycle. Inherited deficiency of this enzyme results in argininemia, an autosomal recessive disorder characterized by hyperammonemia.
Specifications

Specifications

Arginase 1
Polyclonal
Unconjugated
ARG1
Liquid
Rabbit
IgG
1 mg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
513608
ELISA, Immunomicroscopy, Western Blot
10 mg/mL
0.02M potassium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% sodium azide; pH 7.2
Q2KJ64
ARG1
Arginase from bovine liver.
IgG fraction
RUO
Antibody
Polyclonal
Bovine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit