Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIX1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524282

Catalog No. PIPA524282

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The specific function of this protein remains unknown.
Specifications

Specifications

LIX1
Polyclonal
PBS with 0.09% sodium azide
Q8N485, Q6P566
C5orf11, limb expression 1, protein limb expression 1 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
167410, 66643
Western Blot
Unconjugated
LIX1
Liquid
LIX1
KLH conjugated synthetic peptide between 221-250 amino acids from the C-terminal region of human LIX1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit