Learn More
lethal giant larvae homolog 2 (Drosophila), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human LLGL2 protein.
Supplier: Abnova Corporation H00003993D01P
Description
The lethal (2) giant larvae protein of Drosophila plays a role in asymmetric cell division, epithelial cell polarity, and cell migration. This human gene encodes a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MRRFLRPGHDPVRERLKRDLFQFNKTVEHGFPHQPSALGYSPSLRILAIGTRSGAIKLYGAPGVEFMGLHQENNAVTQIHLLPGQCQLVTLLDDNSLHLWSLKVKGGASELQEDESFTLRGPPGAAPSATQITVVLPHSSCELLYLGTESGNVFVVQLPAFRALEDRTISSDAVLQRLPEEARHRRVFEMVEALQEHPRDPNQILIGYSRGLVVIWDLQGSRVLYHFLSSQQLENIWWQRDGRLLVSCHSDGSYCQWPVSSEAQQPEPLRSLVPYGPFPCKAITRILWLTTRQGLPFTIFQGGMPRASYGDRHCISVIHDGQQTAFDFTSRVIGFTVLTEADPAATFDDPYALVVLAEEELVVIDLQTAGWPPVQLPYLASLHCSAITCSHHVSNIPLKLWERIIAAGSRQNAHFSTMEWPIDGGTSLTPAPPQRDLLLTGHEDGTVRFWDASGVCLRLLYKLSTVRVFLTDTDPNENLSAQGEDEWPPLRKVGSFDPYSDDPRLGIQKIFLCKYSGYLAVAGTAGQVLVLELNDEAAEQAVEQVEADLLQDQEGYRWKGHERLAARSGPVRFEPGFQPFVLVQCQPPAVVTSLALHSEWRLVAFGTSHGFGLFDHQQRRQVFVKCTLHPSDQLALEGPLSRVKSLKKSLRQSFRRMRRSRVSSRKRHPAGPPGEAQEGSAKAERPGLQNMELAPVQRKIEARSAEDSFTGFVRTLYFADTYLKDSSRHCPSLWAGTNGGTIYAFSLRVPPAERRMDESVRAEQAKEIQLMHRAPVVGILVLDGHSVPLPEPLEVAHDLSKSPDMQGSHQLLVVSEEQFKVFTLPKVSAKLKLKLTALEGSRVRRVSVAHFGSRRAEDYGEHHLAVLTNLGDIQVVSLPLLKPQVRYSCIRREDVSGIASCVFTKYGQGFYLISPSEFERFSLSTKWLVEPRCLVDSAETKNHRPGNGAGPKKAPSRARNSGTQSDGEEKQPGLVMERALLSDERVLKEIQSTLEGDRGSGNWRSHRAAVGCSLSNGGAESpecifications
lethal giant larvae homolog 2 (Drosophila) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human LLGL2 protein. | |
LLGL2 | |
HGL/LGL2 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC064994.1 | |
LLGL2 | |
LLGL2 (AAH64994.1, 1 a.a. ∼ 1020 a.a) full-length human protein. | |
100 μg | |
Cell Cycle | |
3993 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.