Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMAN1, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Supplier:  Thermo Scientific PA127669

Catalog No. PIPA127669

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Mannose-specific lectin. May recognize sugar residues of glycoproteins. glycolipids. or glycosylphosphatidyl inositol anchors and may be involved in the sorting or recycling of proteins. lipids. or both. The LMAN1-MCFD2 complex forms a specific cargo rece

PA1-27669 detects LMAN1 from human samples.PA1-27669 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27494 immunogen is a synthetic peptide corresponding to human ERGIC-53 protein.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMAN1
Polyclonal
LMAN1
ERGIC-53, ERGIC53, F5F8D, FMFD1, MCFD1, MR60, gp58
Chicken
Antigen affinity chromatography
RUO
3998
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Purified
Western Blot
Unconjugated
P49257
LMAN1
Synthetic peptide corresponding to human ERGIC-53 protein.
50 μg
Primary
Human
Antibody
IgY
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit