Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMF2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524384

Catalog No. PIPA524384

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.
Specifications

Specifications

LMF2
Polyclonal
PBS with 0.09% sodium azide
Q9BU23
TMEM112B, TMEM153, transmembrane protein 112B, transmembrane protein 153
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
91289
Flow Cytometry, Western Blot
Unconjugated
LMF2
Liquid
LMF2
KLH conjugated synthetic peptide between 535-564 amino acids from the C-terminal region of human LMF2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit