Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LMO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA527493
Description
Recommended positive controls: 293T, mouse brain, rat brain. Predicted reactivity: Mouse (98%), Rat (99%), Zebrafish (98%), Xenopus laevis (98%), Pig (96%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LMO1 Polyclonal antibody specifically detects LMO1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LMO1 | |
Polyclonal | |
Unconjugated | |
LMO1 | |
cb524; Cysteine-rich protein TTG-1; Dat1; dopamine-inducible LIM-domain transcription factor DAT1; etID41074.7; LIM domain only 1; LIM domain only 1 (rhombotin 1); LIM domain only 3; LIM domain only protein 1; LIM domain only protein 3; LIM-only 1; Lmo1; LMO-1; lmo1b; Lmo3; LMO-3; neuronal-specific transcription factor DAT1; RBTN1; Rbtn-1; RHOM1; rhombotin 1; rhombotin-1; T-cell translocation gene 1; T-cell translocation protein 1; transcription factor; Ttg1; Unknown (protein for MGC:142594); wu:fb76g02; zgc:109794 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
109594, 245979, 4004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.87 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P25800, Q924W9 | |
LMO1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 156 of Human LMO1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction