Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMOD2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523868

Catalog No. PIPA523868


Add to cart

Description

Description

This antibody is predicted to react with rat based on sequence homology.

Specifications

Specifications

LMOD2
Polyclonal
PBS with 0.09% sodium azide
Q6P5Q4, Q3UHZ5
C-LMOD, cardiac leiomodin, leiomodin-2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
442721, 93677
Immunohistochemistry, Western Blot
Unconjugated
LMOD2
Liquid
LMOD2
KLH conjugated synthetic peptide between 60-88 amino acids from the N-terminal region of human LMOD2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit