Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMX1A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511474

Catalog No. PIPA511474

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Insulin is produced exclusively by the beta cells in the islets of Langerhans in the pancreas. The level and beta-cell specificity of insulin gene expression are regulated by a set of nuclear genes that bind to specific sequences within the promoter of the insulin gene (INS; MIM 176730) and interact with RNA polymerase to activate or repress transcription. LMX1 is a homeodomain protein that binds an A/T-rich sequence in the insulin promoter and stimulates transcription of insulin.
Specifications

Specifications

Lmx1a
Polyclonal
PBS with 0.09% sodium azide
Q8TE12, Q9JKU8
LIM homeobox transcription factor 1 alpha;LIM/homeobox protein LMX1A;LIM/homeobox protein 1.1;LMX-1.1; Lmx1a
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
110648, 4009
Western Blot
Unconjugated
LMX1A
Liquid
LMX1A
KLH conjugated synthetic peptide between 130-159 amino acids from the central region of human Lmx1a
Protein G
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit