Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

von Willebrand factor A domain containing 5A, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human LOH11CR2A protein.

Manufacturer:  Abnova Corporation H00004013B01

Catalog No. 89-005-707


Add to cart

Description

Description

Sequence: MVHFCGLLTLHREPVPLKSISVSVNIYEFVAGVSATLNYENEEKVPLEAFFVFPMDEDSAVYSFEALVDGKKIVAELQDKMKARTNYEKAISQGHQAFLLEGDSSSRDVFSCNVGNLQPGSKAAVTLKYVQELPLEADGALRFVLPAVLNPRYQFSGSSKDSCLNVKTPIVPVEDLPYTLSMVATIDSQHGIEKVQSNCPLSPTEYLGEDKTSAQVSLAAGHKFDRDVELLIYYNEVHTPSVVLEMGMPNMKPGHLMGDPSAMVSFYPNIPEDQPSNTCGEFIFLMDRSGSMQSPMSSQDTSQLRIQAAKETLILLLKSLPIGCYFNIYGFGSSYEACFPESVKYTQQTMEEALGRVKLMQADLGGTEILAPLQNIYRGPSIPGHPLQLFVFTDGEVTDTFSVIKEVRINRQKHRCFSFGIGEGTSTSLIKGIARASGGTSEFITGKDRMQSKALRTLKRSLQPVVEDVSLSWHLPPGLSAKMLSPEQTVIFRGQRLISYAQLTGRMPAAETTGEVCLKYTLQGKTFEDKVTFPLQPKPDVNLTIHRLAAKSLLQTKDMGLRETPASDKKDALNLSLESGVISSFTAFIAINKELNKPVQGPLAHRDVPRPILLGASAPLKIKCQSGFRKALHSDRPPSASQPRGELMCYKAKTFQMDDYSLCGLISHKDQHSPGFGENHLVQLIYHQNANGSWDLNEDLAKILGMSLEEIMAAQPAELVDSSGWATILAVIWLHSNGKDLKCEWELLERKAVAWMRAHAGSTMPSVVKAAITFLKSSVDPAIFAF
Specifications

Specifications

von Willebrand factor A domain containing 5A
Unconjugated
No additive
NM_014622.4
BCSC-1|BCSC1|LOH11CR2A
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human LOH11CR2A protein.
VWA5A
Serum
VWA5A
LOH11CR2A (NP_055437.2, 1 a.a. ~ 786 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
4013
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit