Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LONP2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526630

Catalog No. PIPA526630


Add to cart

Description

Description

LONP2 is probable serine protease which may be involved in peroxisome biogenesis.
Specifications

Specifications

LONP2
Polyclonal
PBS with 0.09% sodium azide
Q86WA8, Q9DBN5
LONP, LONPL, lon protease 2, lon protease homolog 2, peroxisomal, lon protease-like protein 2, peroxisomal LON protease like, peroxisomal Lon protease homolog 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
66887, 83752
Western Blot
Unconjugated
LONP2
Liquid
LONP2
KLH conjugated synthetic peptide between 104-132 amino acids from the N-terminal region of human LONP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit