Learn More
anti-LOX, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NB11059729SS
 View more versions of this product
Catalog No. NB11059729T
Add to cart
Description
Description
LOX Polyclonal antibody specifically detects LOX in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
LOX
Unconjugated
Whole antisera with 0.1% Sodium Azide
Antisera
LOX
Two synthetic peptides (within residues 150-200 and 275-325) of the rat LOX protein. [UniProt# P16636]
33 kDa
0.025 ml
Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary
4015
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Western Blot 1:4000, Immunohistochemistry 1:100 - 1:250, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:100 - 1:250, Immunohistochemistry-Frozen
P16636
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Rabbit
IgG
Unpurified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat, Primate, Rabbit