Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LOX propeptide, Alexa Fluor 700, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130012AF700

Catalog No. NBP130012Z


Add to cart

Description

Description

LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

LOX propeptide
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
LOX
A synthetic peptide made to an internal region of mouse LOX propeptide (within residues 135-155). [Swiss-Prot P28301]
50 kDa
0.1 ml
Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary
4015
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
P28300
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human and mouse. Immunogen has 100% homology with rat.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only