Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-LOX propeptide, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP130327H

Catalog No. NBP130327H


Add to cart

Description

Description

LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Rat samples. It is validated for Western Blot.
Specifications

Specifications

LOX propeptide
HRP
PBS with No Preservative
Affinity Purified
LOX
Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot# P28300]
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
This antibody recognizes the glycosylated propeptide form (~35 kDa) and the proenzyme form (~50 kDa).
Western Blot
Western Blot
P28300
lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Rabbit
IgG
0.1 ml
Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction
Primary
4015
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only