Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523001

Catalog No. PIPA523001


Add to cart

Description

Description

The target sequence has 100% sequence homology with rat.

Lysyl oxidase (LOX) is an extracellular enzyme that is critical for the normal biosynthesis of collagens and elastin. LOX is synthesized as a 50 kDa precursor, secreted into the extracellular matrix and subsequently proteolytically processed to a 30 kDa active enzyme and an 18 kDa propeptide. LOX also reverts ras-mediated transformation and its expression is down-regulated in human cancers. Most recently, the lysyl oxidase pro-peptide (LOPP) has been associated with tumor suppression, as it inhibits ras-dependent cellular transformation.
Specifications

Specifications

LOX
Polyclonal
Unconjugated
LOX
Liquid
LOX
A synthetic peptide made to an internal region of mouse LOX propeptide (within residues 135-155).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.1% sodium azide
P28300, P28301
protein-lysine 6-oxidase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
16948, 4015
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit