lysyl oxidase-like 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against a full-length human LOXL1 protein.

Manufacturer: Abnova Corporation H00004016D01P

Catalog No. 89-017-819

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Applications Western Blotting
    Formulation In 1X PBS, pH 7.4
    Gene LOXL1
    Gene Accession No. NM_005576.2
    Gene Alias LOL, LOXL
    Gene Symbols LOXL1
    Species Reactivity Human
    Storage Requirements Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
    Gene ID (Entrez) 4016
    Description lysyl oxidase-like 1
    Antigen lysyl oxidase-like 1
    Format Affinity Purified
    Host Species Rabbit
    Immunogen LOXL1 (NP_005567.2, 1 a.a. ∽574 a.a) full-length human protein.
    Purification Method Protein A column
    Quantity 100μg
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Conjugate Unlabeled
    Cross Reactivity Human

    This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family. [provided by RefSeq

    Sequence: MALARGSRQLGALVWGACLCVLVHGQQAQPGQGSDPARWRQLIQWENNGQVYSLLNSGSEYVPAGPQRSESSSRVLLAGAPQAQQRRSHGSPRRRQAPSLPLPGRVGSDTVRGQARHPFGFGQVPDNWREVAVGDSTGMARARTSVSQQRHGGSASSVSASAFASTYRQQPSYPQQFPYPQAPFVSQYENYDPASRTYDQGFVYYRPAGGGVGAGAAAVASAGVIYPYQPRARYEEYGGGEELPEYPPQGFYPAPERPYVPPPPPPPDGLDRRYSHSLYSEGTPGFEQAYPDPGPEAAQAHGGDPRLGWYPPYANPPPEAYGPPRALEPPYLPVRSSDTPPPGGERNGAQQGRLSVGSVYRPNQNGRGLPDLVPDPNYVQASTYVQRAHLYSLRCAAEEKCLASTAYAPEATDYDVRVLLRFPQRVKNQGTADFLPNRPRHTWEWHSCHQHYHSMDEFSHYDLLDAATGKKVAEGHKASFCLEDSTCDFGNLKRYACTSHTQGLSPGCYDTYNADIDCQWIDITDVQPGNYILKVHVNPKYIVLESDFTNNVVRCNIHYTGRYVSATNCKIVQS

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit