Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPAR1/LPA1/EDG-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
$204.00 - $482.50
Specifications
Antigen | LPAR1/LPA1/EDG-2 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5 - 2.0 ug/ml, Simple Western 1:200, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:100 -1:500, Immunohistochemistry-Frozen 1:100-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP103363SS
|
Novus Biologicals
NBP103363SS |
0.025 mL |
Each for $204.00
|
|
|||||
NBP103363
|
Novus Biologicals
NBP103363 |
0.1 mL |
Each for $482.50
|
|
|||||
Description
LPAR1/LPA1/EDG-2 Polyclonal specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
LPAR1/LPA1/EDG-2 | |
Western Blot 0.5 - 2.0 ug/ml, Simple Western 1:200, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:100 -1:500, Immunohistochemistry-Frozen 1:100-1:500 | |
Polyclonal | |
Rabbit | |
Apoptosis, GPCR, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 | |
LPAR1 | |
IgG | |
Affinity Purified | |
Human, Chicken and Rat. Predicted to react with sheep, mouse, and bovine based on 100% sequence homology. Positive staining in IHC-Fr/IF with 16 um cryostat sections of embryonic day 6 (E6) chicken brain was reported by a customer. |
1 mg/mL | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Rat, Chicken | |
Q92633 | |
1902 | |
Synthetic peptide made to an internal portion of human EDG2 (within residues 200-300). [Swiss-Prot# Q92633] | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
43 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title