Learn More
LPIN1 Mouse anti-Human, Clone: 3D9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA521419
Catalog No. PIMA521419
Add to cart
Description
Description
Peptide Sequence: EPFYAAFGNR PADVYSYKQV GVSLNRIFTV NPKGELVQEH AKTNISSYVR LCEVVDHVFP LLKRSHSSDF PCSDTFSNFT FWREPLPPFE NQDIHSASA
This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism.
Specifications
Specifications
LPIN1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q14693
LPIN1, PAP1, Lipin 1, fatty liver dystrophy protein, phosphatidate phosphatase LPIN1
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23175
ELISA, Western Blot
3D9
Unconjugated
LPIN1
Liquid
LPIN1
LPIN1 (NP_663731.1, 792 a.a. ∽ 890 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human